Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp90 Mouse, Unlabeled, Clone: 68, BD

Mouse Monoclonal Antibody

$232.00 - $458.00

Specifications

Antigen Hsp90
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610418
View Documents Promotion Details
BD Biosciences
610418
50μg
Each for $232.00
Add to cart
 
BDB610419
View Documents Promotion Details
BD Biosciences
610419
150μg
Each for $458.00
Add to cart
 
Description

Description

The 90kDa heat shock protein (Hsp90), a highly conserved stress-induced protein, is abundantly expressed in most tissues under nonstress conditions and is required for eukaryotic cell viability. Hsp90 interacts with a number of signaling molecules, other heat shock proteins, and cytoskeletal proteins. However, the function of Hsp90 in these complexes remains unclear. It is thought that Hsp90 plays a common role in a number of diverse signaling systems due to its physical association with v-Src family tyrosine kinases, steroid hormone receptors, the Raf serine/threonine kinase, the basic-helix-loop-helix dioxin receptor, and the βγ subunits of G proteins.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Hsp90
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Heat Shock Protein-90
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
68
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit