Learn More
heat shock 70kDa protein 1-like, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a partial recombinant HSPA1L.
Manufacturer: Abnova Corporation H00003305A01
Description
This gene encodes a 70kDa heat shock protein. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which also encode isoforms of the 70kDa heat shock protein. [provided by RefSeq
Sequence: KGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVDSpecifications
heat shock 70kDa protein 1-like | |
Unconjugated | |
50% glycerol | |
NM_005527 | |
HSP70-1L|HSP70-HOM|HSP70T|hum70t | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
3305 |
ELISA, Western Blot | |
Mouse polyclonal antibody raised against a partial recombinant HSPA1L. | |
HSPA1L | |
Serum | |
HSPA1L | |
HSPA1L (NP_005518, 561 a.a. ~ 641 a.a) partial recombinant protein with GST tag. | |
RUO | |
Y | |
Polyclonal | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.