Learn More
HSPA2 Mouse anti-Human, Clone: 3H7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519150
Catalog No. PIMA519150
Add to cart
Description
Description
Peptide Sequence: MSARGPAIGI DLGTTYSCVG VFQHGKVEII ANDQGNRTTP SYVAFTDTER LIGDAAKNQV AMNPTNTIFD AKRLIGRKFG DATVQSDMKH WPFRVVSEGG KPKVQVEYKG ETKTFFPEEI SSMVLTKMKE IAEAYLGGKV HSAVITVPAY FNDSQRQATK DAGTITGLNV LRIINEPTAA AIAYGLDKKG CAGGEKNVLI FDLGGGTFDV SILTIEDGIF EVKSTAGDTH LGGEDFDNRM VSHLAEEFKR KHKKDIGPNK RAVRRLRTAC ERAKRTLSSS TQASIEIDSL YEGVDFYTSI TRARFEELNA DLFRGTLEPV EKALRDAKLD KGQIQEIVLV GGSTRIPKIQ KLLQDFFNGK ELNKSINPDE AVAYGAAVQA AILIGDKSEN VQDLLLLDVT PLSLGIETAG GVMTPLIKRN TTIPTKQTQT FTTYSDNQSS VLVQVYEGER AMTKDNNLLG KFDLTGIPPA PRGVPQIEVT FDIDANGILN VTAADKSTGK ENKITITNDK GRLSKDDIDR MVQEAERYKS EDEANRDRVA AKNALESYTY NIKQTVEDEK LRGKISEQDK NKILDKCQEV INWLDRNQMA EKDEYEHKQK ELERVCNPII SKLYQGGPGG GSGGGGSGAS GGPTIEEVD
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Specifications
Specifications
HSPA2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P54652
Heat shock-related 70 kDa protein 2; Heat shock 70 kDa protein 2; HSPA2; HSP70-3; HSP70-2
Mouse
IgG3, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3306
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
3H7
Unconjugated
HSPA2
Liquid
HSPA2
HSPA2 (AAH01752, 1 a.a. ∽ 639 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human