Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

HSPA9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523669

Catalog No. PIPA523669

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the heat shock protein 70 gene family. The encoded protein is primarily localized to the mitochondria but is also found in the endoplasmic reticulum, plasma membrane and cytoplasmic vesicles. This protein is a heat-shock cognate protein. This protein plays a role in cell proliferation, stress response and maintece of the mitochondria. A pseudogene of this gene is found on chromosome 2.
Specifications

Specifications

HSPA9
Polyclonal
PBS with 0.09% sodium azide
P38646
CSA; GRP-75; GRP75; HSPA9B; MOT; MOT2; MTHSP75; PBP74; 75 kDa glucose-regulated protein; heat shock 70kD protein 9B; mortalin; perinuclear; mortalin-2; p66-mortalin; peptide-binding protein 74; stress-70 protein; mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3313
Western Blot
Unconjugated
HSPA9
Liquid
HSPA9
KLH conjugated synthetic peptide between 273-301 amino acids from the central region of human HSPA9
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit