Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP20 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526634

Catalog No. PIPA526634

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HSPB6 is associated with actin (see MIM 102540) and modulates smooth muscle relaxation.
Specifications

Specifications

HSP20
Polyclonal
PBS with 0.09% sodium azide
O14558, Q5EBG6
Hsp20; heat shock 20 kDa-like protein p20; heat shock protein beta-6; HSPB6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
126393, 243912
Western Blot
Unconjugated
HSPB6
Liquid
HSPB6
KLH conjugated synthetic peptide between 91-120 amino acids from the central region of human HSPB6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit