Learn More
HSPH1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524619
Catalog No. PIPA524619
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is predicted to react with bovine based on sequence homology.
HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.
Specifications
Specifications
HSPH1
Polyclonal
Unconjugated
HSPH1
Liquid
HSPH1
KLH conjugated synthetic peptide between 549-579 amino acids from the central region of human HSPH1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q92598
Heat shock protein 105 kDa; Heat shock 110 kDa protein; Antigen NY-CO-25; HSP105; HSP110; KIAA0201
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10808