Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-HU A+G+M (H+L) AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300289
Affinity purified polyclonal antibody to Human IgA, IgG, and IgM, both heavy and light chains (whole IgA, IgG, and IgM) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-668-75