Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-HU A+G+M (H+L) AB MSA HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200278
Affinity purified polyclonal secondary antibody against human IgA, IgG, and IgM, both heavy and light chains (whole IgA, IgM, and IgG) made in Goat and labeled with horseradish peroxidase.
Catalog No. 50-669-33