Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-HU IGA+IGG+IGM(H+L)AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200331
Affinity purified polyclonal antibody to Human IgA, IgG, and IgM, heavy and light chains (whole IgA, IgG, and IgM) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-671-96