Learn More
Seracare Life Sciences Inc ANTI-HU IGG (H+L) F(AB )2 CY5
Supplier:  Seracare Life Sciences Inc 52300429
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab')2 fragment, to human IgG heavy and light chains (H+L) made in goat and labeled with Cy5. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-673-18
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.