Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-HUIGG(GAMMA) F(AB )2 FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300441
Affinity purified polyclonal antibody, F(ab)2 fragment, to Human IgG heavy chains (gamma). Made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-52