Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM9 Ectodomain Goat anti-Human, Polyclonal, R&D Systems
Goat Polyclonal Antibody
Manufacturer: R&D Systems AF939
This item is not returnable.
View return policy
Description
ADAM9 Ectodomain Polyclonal antibody AF939 Specifically detects ADAM9 Ectodomain in Human samples. It is validated for ELISA Capture, Immunoprecipitation, Western BlottingSpecifications
ADAM9 | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q13443 | |
a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMP, MCMPMDC9KIAA0021Mltng, MDC9, Meltrin gamma, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
Goat | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human ADAM9 Ectodomain in ELISAs and Western blots. In sandwich immunoassays, approximately 20% cross-reactivity with recombinant mouse (rm) rmADAM9 is observed and less than 0.05% cross-reactivity with recombinant human (rh) rhADAM8, rhADAM10, rhTACE, rhTIMP-1, rhTIMP-2, rhTIMP-3, rhTIMP-4, rhBACE-1 and rmADAM10 is observed. | |
Human |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
ADAM9 | |
Mouse myeloma cell line NS0-derived recombinant human ADAM9 Ala206-Asp697 Accession # Q13443 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
8754 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title