Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human C1qR1/CD93 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: R&D Systems BAF2379
This item is not returnable.
View return policy
Description
C1qR1/CD93 Polyclonal specifically detects C1qR1/CD93 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
C1qR1/CD93 | |
Polyclonal | |
Biotin | |
Q9NPY3 | |
CD93 | |
Mouse myeloma cell line NS0-derived recombinant human C1q R1/CD93 Ala24-Lys580 Accession # Q9NPY3 | |
50 μg | |
Primary | |
Detects human C1q R1/CD93 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse C1q R1 is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
AA4 Antigen, C1q R1, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 | |
Goat | |
Affinity Purified | |
RUO | |
22918 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction