Learn More
Caspase-10 (FLICE 2) Rabbit anti-Human, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Manufacturer:  R&D Systems AF834
Catalog No. AF834
Add to cart
This item is not returnable. View return policy
Description
Description
Caspase-10 (FLICE 2) Polyclonal antibody AF834 Specifically detects Caspase-10 (FLICE 2) in Human samples. It is validated for Western Blotting
Specifications
Specifications
Caspase-10/FLICE2
LYOPH
Western Blot 1 ug/mL
Affinity Purified
CASP10
KLH-coupled human Caspase-10 (FLICE 2) synthetic peptide KRTVWGAKQISATSLPTAC
Immunogen affinity purified
RUO
Primary
843
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Apoptotic protease Mch-4, CASP10, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, Caspase10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, Mch4, MCH4ALPS2
Rabbit
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse Caspase-10 (FLICE 2) precursor in Western blots. Sensitivity for detection of the small subunit is not known.
Human