Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cathepsin D Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems BAF1014
This item is not returnable.
View return policy
Description
Cathepsin D Polyclonal specifically detects Cathepsin D in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cathepsin D | |
Polyclonal | |
Biotin | |
P07339 | |
CTSD | |
Mouse myeloma cell line NS0-derived recombinant human Cathepsin D Leu21-Leu412 Accession # P07339 | |
50 μg | |
Primary | |
Detects human Cathepsin D in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse Cathepsin D is observed, and less than 1% cross-reactivity with recombinant human (rh) Cathepsin A, rhCathepsin B, rhCathepsin C, rhCathepsin L, and rhCathepsin X/Z/P is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
Goat | |
Affinity Purified | |
RUO | |
1509 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction