Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-Human CD11b/MAC-1; Clone: ICRF44; BD

$195.00 - $395.00

Specifications

Antigen Human CD11b/MAC-1
Clone ICRF44
Format Affinity Purified
Host Species Murine
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB562723
View Documents
BD Biosciences
562723
25 Tests Each for $195.00
N/A
BDB562721
View Documents
BD Biosciences
562721
100 Tests Each for $395.00
N/A
Description

Description

Host Species: Mouse
Clone: ICRF44
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Immunogen: Human monocytes

Applications:

Flow Cytometry

Specifications

Specifications

Human CD11b/MAC-1
Affinity Purified
RUO
Monoclonal
ICRF44
Murine
Human monocytes
Primary
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit