Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Human CD152, Brilliant Violet™ 421, Clone: BNI3; BD
Supplier: BD Biosciences 562743
Description
Host Species: Mouse
Clone: BNI3
Isotype: IgG2a, κ
Cross Reactivity Species: Hu
Immunogen: Human CTLA4 Recombinant Protein
Applications:
Flow Cytometry
Specifications
| Human CD152 | |
| BNI3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| Brilliant Violet 421 | |
| Human CTLA4 Recombinant Protein | |
| 50 Tests | |
| Primary | |
| IgG2a |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction