Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human CD62L; Clone: DREG-56; BD

$195.00 - $395.00

Specifications

Antigen Human CD62L
Clone DREG-56
Format Affinity Purified
Host Species Murine
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB562720
View Documents
BD Biosciences
562720
25 Tests
Each for $195.00
Add to cart
 
BDB562719
View Documents
BD Biosciences
562719
100 Tests
Each for $395.00
Add to cart
 
Description

Description

Host Species: Mouse
Clone: DREG-56
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Immunogen: Supernatant from PMA-activated Human Peripheral Blood Leukocytes

Applications:

Flow Cytometry

Specifications

Specifications

Human CD62L
Affinity Purified
RUO
Monoclonal
DREG-56
Murine
Supernatant from PMA-activated Human Peripheral Blood Leukocytes
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit