Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF284
This item is not returnable.
View return policy
Description
Common gamma Chain/IL-2 R gamma Polyclonal specifically detects Common gamma Chain/IL-2 R gamma in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.Specifications
Common gamma Chain/IL-2 R gamma | |
Polyclonal | |
Unconjugated | |
P31785 | |
IL2RG | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Common gamma Chain/IL-2 R gamma Leu23-Asn254 Accession # P31785 | |
250 μg | |
Primary | |
Detects human Common gamma Chain/IL-2 R gamma in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse IL-2 R gamma is observed, less than 10% cross-reactivity with recombinant human (rh) IL-2 R beta is observed, and less than 2% cross-reactivity with rhIL-2 R alpha is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 20-60 ug/mL, CyTOF-ready | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 R gamma, IL-2 receptor subunit gamma, IL2R gamma, IL-2R subunit gamma, IL2RG, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
Goat | |
Affinity Purified | |
RUO | |
3561 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title