Learn More
DC-SIGN/CD209 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF161
Catalog No. AF161
Add to cart
This item is not returnable. View return policy
Description
Description
DC-SIGN/CD209 Polyclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot.
Specifications
Specifications
DC-SIGN/CD209
LYOPH
Western Blot 1 ug/mL
Q9NNX6
CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human DC-SIGN/CD209 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CD209
Mouse myeloma cell line NS0-derived recombinant human DC-SIGN/CD209 Lys62-Ala404 Accession # Q9NNX6
Antigen Affinity-purified
RUO
Primary
30835
Reconstitute at 0.2 mg/mL in sterile PBS.