Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETV6 Sheep anti-Human, Polyclonal, R&D Systems
Sheep Polyclonal Antibody
Manufacturer: R&D Systems AF7945
This item is not returnable.
View return policy
Description
ETV6 Polyclonal antibody AF7945 Specifically detects ETV6 in Human samples. It is validated for Western BlottingSpecifications
ETV6/Tel | |
LYOPH | |
Western Blot 1 ug/mL | |
P41212 | |
ETS translocation variant 6, ets variant 6, ets variant gene 6 (TEL oncogene), ETS-related protein Tel1, Tel, TEL/ABL, TEL1, TELTEL1 oncogene, transcription factor ETV6 | |
Sheep | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human ETV6 in direct ELISAs and Western blots. | |
Human |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
ETV6 | |
E. coli-derived recombinant human ETV6 Ser2-Asp111 Accession # P41212 | |
Immunogen affinity purified | |
RUO | |
Primary | |
2120 | |
Sterile PBS to a final concentration of 0.2 mg/mL. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title