Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FosB/G0S3 Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF2214
This item is not returnable.
View return policy
Description
FosB/G0S3 Polyclonal specifically detects FosB/G0S3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
FosB/G0S3 | |
Polyclonal | |
Unconjugated | |
P53539 | |
FOSB | |
E. coli-derived recombinant human FosB Met1-Leu338 Accession # P53539 | |
100 μg | |
Primary | |
Detects human FosB. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
activator protein 1, AP-1, DKFZp686C0818, FBJ murine osteosarcoma viral oncogene homolog B, G0S3, G0S3G0/G1 switch regulatory protein 3, GOS3, GOSB, MGC42291, oncogene FOS-B, protein fosB | |
Goat | |
Affinity Purified | |
RUO | |
2354 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction