Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone Rabbit anti-Human, Porcine, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP150119

Catalog No. NBP150119


Add to cart

Description

Description

Growth Hormone Polyclonal antibody specifically detects Growth Hormone in Human, Porcine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Growth Hormone
Unconjugated
Diluted whole antisera with 0.02% Sodium Azide
Antisera
GH1
The antibody reacts with the Growth Hormone antigen.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The Growth Hormone antibody can be used for the diagnosis of tumors of the pituitary.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P01241
GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N
Rabbit
IgG
0.25 ml
Biologically Active Proteins
Primary
2688
Human, Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only