Learn More
Pepro Tech Inc Anti-Human HB-EGF, 100ug DFS Item
Manufacturer:  Pepro Tech Inc 500P329100UG
Produced from sera of rabbits immunized with highly pure Recombinant Human HB-EGF. Anti-Human HB-EGF-specific antibody was purified by affinity chromatography employing an immobilized Human HB-EGF matrix.
Catalog No. 50-195-2597