Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F(ab')2-Goat anti-Human Kappa Light Chain Secondary Antibody, HRP, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Supplier: Thermo Scientific PA174407
Description
This antibody has been cross-absorbed with pooled human myeloma proteins with lambda light chains. Purified IgG was prepared from whole serum by affinity chromatography, and F(ab)2 fragments were prepared by pepsin digestion. Goat F(ab')2 anti Human Kappa Light Chain antibody recognizes human kappa light chains. This antibody has been cross absorbed with pooled human myeloma proteins with lambda light chains.
Human Kappa Light Chain Polyclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Human Kappa Light Chain | |
Polyclonal | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.4 | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
HRP | |
Iglc | |
Human kappa light chains. | |
1 mL | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction