Learn More
IgA Mouse anti-Human, Invitrogen™
Mouse Monoclonal Secondary Antibody
Manufacturer:  Invitrogen LFMA0165
Catalog No. LFMA0165
Add to cart
Description
Description
A suggested positive control for this product is human plasma.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Human IgA
Monoclonal
1 mg/mL
HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide
Mouse
IgG1, kappa
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Secondary
Human
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
47C12
Unconjugated
Liquid
Protein purified from Human plasma
Protein G
RUO
Secondary Antibody
Monoclonal