Learn More
anti-Human IgA, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP142088
Catalog No. NBP142088
Add to cart
Description
Description
Human IgA Polyclonal antibody specifically detects Human IgA in Human samples. It is validated for ELISA, Dot Blot.
Specifications
Specifications
Human IgA
Unconjugated
50% glycerol, 0.1 M NaCl, 0.01 M sodium phosphate at pH 7.5 with No Preservative
Rabbit
IgG
1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Dot Blot, ELISA
ELISA 1:100, Dot Blot 1:100
Purified
The antibody reacts with the Human IgA antigen.
IgG purified
RUO
Primary
Antibodies were raised in rabbit to human IgA. From the crude polyclonal the cross reactive antibodies were extracted by incubation with Sepharose-bound human IgG and IgM. Specific antibodies were absorpted by incubation with Sepha¡rose-bound human IgA. Specific antibodies were eluted by acidic buffer at pH 2.5 followed by neutralisation and dialysis. After repeated binding with immobilized human IgA a minimum of 65% protein bound.