Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-1 Goat anti-Human, Polyclonal, R&D Systems
Goat Polyclonal Antibody
Manufacturer: R&D Systems AF871
This item is not returnable.
View return policy
Description
IGFBP-1 Polyclonal antibody AF871 Specifically detects IGFBP-1 in Human samples. It is validated for Simple Western, Western BlottingSpecifications
IGFBP-1 | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
P08833 | |
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP | |
Goat | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human IGFBP-1 in direct ELISAs and Western blots. In these formats, less than 2% cross-reactivity with recombinant human (rh) IGFBP-2, rhIGFBP-3, rhIGFBP-4, and rhIGFBP-5 is observed. | |
Human |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
IGFBP1 | |
E. coli-derived recombinant human IGFBP-1 Ala26-Asn259 Accession # P08833 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
3484 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title