Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-HUMAN IGG (FC) AB AP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200304
Affinity purified polyclonal secondary antibody against the Fc fragment of human IgG (whole IgG) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-03