Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-HUMAN IGG (GAMMA) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200456
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat and labeled with horseradish peroxidase. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-84