Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-HUMAN IGG (H+L) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200277
Affinity purified polyclonal antibody to Human IgG, both heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.
Catalog No. 50-669-32