Learn More
anti-Human IgG, HRP, Isotype Control, Novus Biologicals™
Human Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197066
Catalog No. NBP197066
Add to cart
Description
Description
IgG antibody specifically detects IgG in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
IgG
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate
immunoglobulin heavy constant gamma 1 (G1m marker)
Human
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
HRP
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Human IgG and anti-Human Serum.
Human