Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human IgG3, Alexa Fluor 546, Isotype Control, Novus Biologicals™

Human Polyclonal Antibody

Manufacturer:  Novus Biologicals DDXCH03A546100

Catalog No. DDXCH03A546


Add to cart

Description

Description

IgG3 Polyclonal antibody specifically detects IgG3 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface), Flow (Intracellular), Immunofluorescence.
Specifications

Specifications

IgG3
0.5mg/mL
Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500
Ion Exchange Chromatography
IGHG3
IgG3
0.1 mg
Immunology
Isotype Control
Polyclonal
Human
Flow Cytometry, Immunofluorescence
Alexa Fluor 546
PBS, 50% glycerol. with No Preservative
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Human
Ion exchange chromatography
RUO
Store at -20C. Avoid freeze-thaw cycles.
Primary
3502
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only