Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGM (MU) AB HRP
Supplier: Seracare Life Sciences Inc 52200328
Affinity purified polyclonal antibody to Human IgM heavy chains (gamma) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable.
View return policy