Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-37/IL-1F7 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1975
This item is not returnable.
View return policy
Description
IL-37/IL-1F7 Polyclonal specifically detects IL-37/IL-1F7 in Human samples. It is validated for Western Blot.Specifications
IL-37/IL-1F7 | |
Polyclonal | |
Unconjugated | |
Q9NZH6-2 | |
IL37 | |
E. coli-derived recombinant human IL-37/IL-1F7 Lys27-Asp192 Accession # Q9NZH6-2 | |
100 μg | |
Primary | |
Detects human IL-37/IL-1F7 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) FIL1 delta , rhFIL1 epsilon , rhFIL1 eta, rhIL-1 alpha , rhIL-1 beta , rhIL-18, and rhIL-1 H1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
FIL1, FIL1 zeta, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, IL37, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
Goat | |
Affinity Purified | |
RUO | |
27178 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title