Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22R alpha 1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF2770
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for Neutralization.Specifications
IL-22R alpha 1 | |
Polyclonal | |
Unconjugated | |
Q8N6P7 | |
IL22RA1 | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1 Pro18-Thr228 Accession # Q8N6P7 | |
100 μg | |
Primary | |
Detects human IL-22 R alpha 1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human IL-20 R beta is observed. | |
Human | |
IgG |
Neutralization | |
LYOPH | |
Neutralization 0.2-1 ug/mL | |
CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
Goat | |
Affinity Purified | |
RUO | |
58985 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title