Learn More
IL-29/IFN-lambda 1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
Manufacturer:  R&D Systems AF1598
Catalog No. AF1598
Add to cart
This item is not returnable. View return policy
Description
Description
IL-29/IFN-lambda 1 Polyclonal specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
Specifications
IL-29/IFN-lambda 1
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.2-1.0 ug/mL
Q8IU54
cytokine Zcyto21, IFNL1, IFN-lambda 1, IFN-lambda-1, IL29, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-29/IFN-lambda 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) IL-28A and rhIL-28B is observed.
Human
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IFNL1
Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200 Accession # Q8IU54
Immunogen affinity purified
RUO
Primary
282618
Reconstitute at 0.2 mg/mL in sterile PBS.