Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-36 gamma/IL-1F9 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems BAF2320
This item is not returnable.
View return policy
Description
IL-36 gamma/IL-1F9 Polyclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot.Specifications
IL-36 gamma/IL-1F9 | |
Polyclonal | |
Biotin | |
Q9NZH8 | |
IL36G | |
E. coli-derived recombinant human IL-36 gamma/IL-1F9 Met1-Asp169 Accession # Q9NZH8 | |
50 μg | |
Primary | |
Detects human IL-36 gamma/IL-1F9 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) IL-36 alpha is observed, and less than 1% cross-reactivity with rhIL-1 alpha , rhIL-1 beta , rhIL-18, rhIL-36Ra, rhIL-37, and rhIL-36 beta is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
IL-1 epsilon, IL-1 H1, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, IL36 gamma, IL36G, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma | |
Goat | |
Affinity Purified | |
RUO | |
56300 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction