Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Supplier: R&D Systems AF209
This item is not returnable.
View return policy
Description
IL-9 Polyclonal specifically detects IL-9 in Human samples. It is validated for Western Blot, Neutralization.Specifications
IL-9 | |
Polyclonal | |
Unconjugated | |
P15248 | |
IL9 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-9 Gln19-Ile144 Accession # P15248 | |
100 μg | |
Primary | |
Detects human IL-9 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant mouse IL-9 and recombinant rat IL-9 is observed. | |
Human | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 2-5 ug/mL | |
Cytokine P40, HP40, IL9, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
Goat | |
Affinity Purified | |
RUO | |
3578 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title