Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP4K5 Sheep anti-Human, Polyclonal, R&D Systems
Sheep Polyclonal Antibody
Manufacturer: R&D Systems AF8007
This item is not returnable.
View return policy
Description
MAP4K5 Polyclonal antibody AF8007 Specifically detects MAP4K5 in Human samples. It is validated for Western BlottingSpecifications
MAP4K5 | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9Y4K4 | |
EC 2.7.11, EC 2.7.11.1, GCKR, germinal center kinase-related, KHS1, KHSMEKKK 5, Kinase homologous to SPS1/STE20, MAPK/ERK kinase kinase kinase 5, MAPKKKK5, MEK kinase kinase 5, mitogen-activated protein kinase kinase kinase kinase 5 | |
Sheep | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human MAP4K5 in direct ELISAs and Western blots. | |
Human |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
MAP4K5 | |
E. coli-derived recombinant human MAP4K5 Lys395-Asn484 (Asn473Lys) Accession # Q9Y4K4 | |
Immunogen affinity purified | |
RUO | |
Primary | |
11183 | |
Sterile PBS to a final concentration of 0.2 mg/mL. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title