Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 zeta Goat anti-Human, Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2669
This item is not returnable.
View return policy
Description
14-3-3 zeta Polyclonal specifically detects 14-3-3 zeta in Human, Mouse samples. It is validated for Western Blot.Specifications
14-3-3 zeta | |
Polyclonal | |
Unconjugated | |
P63104 | |
YWHAZ | |
E. coli-derived recombinant human 14-3-3 zeta Met1-Asn245 Accession # P63104 | |
100 μg | |
Primary | |
Detects human, mouse, and rat 14-3-3 zeta in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human 14-3-3 beta, epsilon, eta, gamma, or sigma is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.25 ug/mL | |
1433 zeta, KCIP-1, KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, YWHAZ, zeta polypeptide | |
Goat | |
Affinity Purified | |
RUO | |
7534 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title