Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Human/Mouse Bax, Polyclonal, R&D Systems
Rabbit Polyclonal Antibody
Manufacturer: R&D Systems AF820
This item is not returnable.
View return policy
Description
Host Species: Rabbit
Species Reactivity:Human, Mouse
Immunogen:KLH-coupled human Bax synthetic peptide, GPTSSEQIMKTGALLLQGFIQDC
Applications:
Immunohistochemistry, Western Blotting
Specifications
Bax | |
LYOPH | |
Western Blot 0.3 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 1-15 ug/mL, Knockout Validated | |
Affinity Purified | |
BAX | |
KLH-coupled human Bax synthetic peptide | |
Immunogen affinity purified | |
RUO | |
Primary | |
581 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Immunocytochemistry, Immunohistochemistry, KnockDown, Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
Rabbit | |
IgG | |
30 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human and mouse Bax. | |
Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title