Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Bcl-2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF810
This item is not returnable.
View return policy
Description
Bcl-2 Polyclonal specifically detects Bcl-2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.Specifications
Bcl-2 | |
Polyclonal | |
Unconjugated | |
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl2, Bcl-2 | |
Goat | |
Affinity Purified | |
RUO | |
596 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 1 ug/10^6 cells | |
BCL2 | |
E. coli-derived recombinant mouse Bcl-2 | |
100 μg | |
Primary | |
Detects human and mouse Bcl-2 in Western blots. | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction