Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-x Rabbit anti-Human, Mouse, Polyclonal, R&D Systems
Rabbit Polyclonal Antibody
Manufacturer: R&D Systems AF800
This item is not returnable.
View return policy
Description
Bcl-x Polyclonal antibody AF800 Specifically detects Bcl-x in Human, Mouse samples. It is validated for Immunoprecipitation, Simple Western, Western BlottingSpecifications
bcl-x | |
LYOPH | |
Western Blot 0.3 ug/mL, Simple Western 3 ug/mL, Immunoprecipitation 5 ug/10^6 cells | |
Affinity Purified | |
Rabbit | |
IgG | |
50 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human and mouse Bcl-x in Western blots. | |
Human, Mouse |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BCL2L1 | |
KLH-coupled Bcl-x synthetic peptide CSAINGNPSWHLADSPAVNGA | |
Immunogen affinity purified | |
RUO | |
Primary | |
598 | |
Reconstitute at 0.2 mg/mL in sterile PBS. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title