Learn More
Casein Kinase 1 epsilon Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF4567
Catalog No. AF4567
Add to cart
This item is not returnable. View return policy
Description
Description
Casein Kinase 1 epsilon Polyclonal antibody AF4567 Specifically detects Casein Kinase 1 epsilon in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications
Specifications
Casein Kinase 1 epsilon
LYOPH
Western Blot 1 ug/mL
P49674
casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, CSNK1E, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects endogenous human, mouse and rat CK1 epsilon in Western blots. In Western blots, this antibody does not cross-react with recombinant human CK1 alpha , CK1 delta , or CK1 gamma .
Human, Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CSNK1E
E. coli-derived recombinant human CK1 epsilon Met317-Lys416 Accession # P49674
Immunogen affinity purified
RUO
Primary
1454
Reconstitute at 0.2 mg/mL in sterile PBS.