Learn More
CIB1 Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF7557
Catalog No. AF7557
Add to cart
This item is not returnable. View return policy
Description
Description
CIB1 Polyclonal antibody AF7557 Specifically detects CIB1 in Human, Mouse, Rat samples. It is validated for Western Blotting
Specifications
Specifications
CIB1
LYOPH
Western Blot 1 ug/mL
Q99828
calcium and integrin binding 1 (calmyrin), Calcium- and integrin-binding protein, calmyrin, CIBcalcium and integrin binding protein, CIBP, DNA-dependent protein kinase interacting protein, DNA-PKcs-interacting protein, Kinase-interacting protein, KIP, KIP1, KIPcalcium and integrin binding, protein kinase interacting protein, PRKDCIP, SIP2-28, SIP2-28calcium and integrin-binding protein 1, Snk interacting protein 2-28, SNK-interacting protein 2-28
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human, mouse, and rat CIB1 in Western blots.
Human, Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CIB1
E. coli-derived recombinant human CIB1 Gly2-Leu191 Accession # Q99828
Immunogen affinity purified
RUO
Primary
10519
Sterile PBS to a final concentration of 0.2 mg/mL.