Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human/Mouse/Rat NMDA R, NR2B Subunit Affinity Purified, Polyclonal, R&D Systems™

Rabbit Polyclonal Antibody

Manufacturer:  R&D Systems PPS013

Catalog No. PPS013


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Rabbit
Species Reactivity:Human, Mouse, Rat
Immunogen:Fusion protein from the C-terminus of the NR2B-subunit of rat NMDA Receptor

Applications:

Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

NMDA R, NR2B Subunit
Unconjugated
10 μg antibody per vial; lyophilized in 5 mM ammonium bicarbonate. with No Preservative
GRIN2B
Fusion protein from the C-terminus of the NR2B-subunit of rat NMDA Receptor
Antigen Affinity-purified
RUO
Primary
2902
This antibody should be reconstituted in 50 μL phosphate buffered saline (137 mM NaCl, 7.5 mM Na2HPO4, 2.7 mM KCl, 1.5 mM KH2PO4, pH 7.4) before use.
Immunohistochemistry, Immunoprecipitation, Western Blot
Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 - 1:2000 dilution, Immunoprecipitation
Affinity Purified
Rabbit
IgG
10 ug
The lyophilized product is stable at ≤ −20°C for at least 1 year. After reconstitution the antibody should be aliquoted and stored at ≤ −20°C.
Polyclonal
Human, mouse, rat ~180 kDa NR2B-subunit of the NMDA Receptor
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit