Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat QDPR Antibody, R&D Systems™
Sheep Polyclonal Antibody
Supplier: R&D Systems AF8038
This item is not returnable.
View return policy
Description
QDPR Polyclonal specifically detects QDPR in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
QDPR | |
Polyclonal | |
Unconjugated | |
P09417 | |
QDPR | |
E. coli-derived recombinant human QDPR Ala2-Phe244 Accession # P09417 | |
100 μg | |
Primary | |
Detects human, mouse, and rat QDPR in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
dhpr, DHPR member 1, HDHPR, PKU2, quinoid dihydropteridine reductaseFLJ42391, SDR33C1 | |
Sheep | |
Affinity Purified | |
RUO | |
5860 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction