Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human/Mouse STAT1 p91, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Manufacturer:  R&D Systems PAFST1

Catalog No. PAFST1


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Species Reactivity:Human, Mouse
Immunogen:E. coli-derived recombinant human STAT1

Applications:

Immunoprecipitation, Western Blotting

Specifications

Specifications

STAT1
LYOPH
Western Blot 0.25-1 ug/mL, Immunoprecipitation 3 ug/10^6 cells, Knockout Validated
Affinity Purified
STAT1
E. coli-derived recombinant human STAT1
Antigen Affinity-purified
RUO
Primary
6772
Sterile PBS to a final concentration of 0.2 mg/mL.
Immunoprecipitation, KnockDown, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse STAT1 p91 in Western blots. In Western blots, less than 1% cross-reactivity with human or mouse STAT1 p84 is observed. Additionally, no cross-reactivity with the other STATs has been detected by immunoprecipitation or Western blotting.
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit