Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human/Mouse STAT4, Polyclonal Affinity Purified, R&D Systems™

Goat Polyclonal Antibody

Manufacturer:  R&D Systems PAFST4

Catalog No. PAFST4


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Species Reactivity:Human, Mouse
Immunogen:Mouse STAT4 synthetic peptide, aa 675-749

Applications:

Immunoprecipitation, Western Blotting

Specifications

Specifications

STAT4
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS. with No Preservative
signal transducer and activator of transcription 4, SLEB11
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse STAT4. No cross-reactivity with the other STATs has been detected by immunoprecipitation or by Western blotting.
Human, Mouse
Immunoprecipitation, Western Blot
Western Blot 0.2 ug/mL, Immunoprecipitation 1 ug/10^6 cells
Affinity Purified
STAT4
Mouse STAT4 synthetic peptide aa 675-749
Immunogen affinity purified
RUO
Primary
6775
Sterile PBS to a final concentration of 0.2 mg/mL.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit